Order Entry
Czech Republic
ContactUsLinkComponent
 
Acetonitrile with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for HPLC
Catalog # 84863.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Acetonitrile with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for HPLC
Catalog # 84863.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    Min. 99.9 %
  • Nominal concentration (TFA; v/v):
    0.0475 to 0.0525 %
  • Evaporation residue :
    Max. 0.0001 %
  • Water:
    Max. 0.005 %
  • Transmittance (195 nm):
    Min.20 %
  • Transmittance (200 nm):
    Min. 50 %
  • Transmittance (245 nm):
    Min. 80 %
  • Transmittance (260 nm):
    Min. 95 %
  • Transmittance (270 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :Packaged under nitrogen.
 :76-05-1