Bis(cyclohexanone)oxalyldihydrazone ≥99.0% for spectrophotometric determination of Cu, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:370-81-0
:
Formula:C₁₄H₂₂N₄O₂
MDL Number:MFCD00001659
:278.36 g/mol
Melting Pt:210…214 °C (lit.)
:M-Clarity™ quality level = MQ100
Frequently Bought Together