Order Entry
Czech Republic
ContactUsLinkComponent
5,5-Dimethyl-1-pyrroline-N-oxide ≥98.0% (by GC) for ESR-spectroscopy, Supelco®
5,5-Dimethyl-1-pyrroline-N-oxide ≥98.0% (by GC) for ESR-spectroscopy, Supelco®
Catalog # 92688-100MG
Supplier:  Merck
CAS Number:  
undefined
5,5-Dimethyl-1-pyrroline-N-oxide ≥98.0% (by GC) for ESR-spectroscopy, Supelco®
Catalog # 92688-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :113.16 g/mol
Storage Temperature:Freezer
CAS Number:3317-61-1
Formula:C₆H₁₁NO
MDL Number:MFCD00005279