Order Entry
Czech Republic
ContactUsLinkComponent
Sodium sulphate 99.0-101.0% (by titrimetric analysis) for HPLC, Supelco®
Sodium sulphate 99.0-101.0% (by titrimetric analysis) for HPLC, Supelco®
Catalog # 80948-250G
Supplier:  Merck
CAS Number:  
Sodium sulphate 99.0-101.0% (by titrimetric analysis) for HPLC, Supelco®
Catalog # 80948-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :142.04 g/mol
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504