Order Entry
Czech Republic
ContactUsLinkComponent
Fluorescein disodium salt 1 mM in CAPS 20 mM solution ≥95.0% (by HPLC) fluorescent dye, Supelco®
Fluorescein disodium salt 1 mM in CAPS 20 mM solution ≥95.0% (by HPLC) fluorescent dye, Supelco®
Catalog # 67884-10ML-F
Supplier:  Merck
CAS Number:  
undefined
Fluorescein disodium salt 1 mM in CAPS 20 mM solution ≥95.0% (by HPLC) fluorescent dye, Supelco®
Catalog # 67884-10ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :376.28 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:518-47-8
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039