Order Entry
Czech Republic
ContactUsLinkComponent
Amberlite® IR120 Hydrogen form 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Amberlite® IR120 Hydrogen form 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Catalog # 10322.
Supplier:  Merck
CAS Number:  
Amberlite® IR120 Hydrogen form 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Catalog # 10322.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
CAS Number:39389-20-3
MDL Number:MFCD00132707