0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Amberlite® XAD®-2
Amberlite® XAD®-2, Pore size: 90 Å mean 20 - 60 mesh ion-exchange resin, Supelco®
Amberlite® XAD®-2, Pore size: 90 Å mean 20 - 60 mesh ion-exchange resin, Supelco®
10357.
:
Merck
:
Amberlite® XAD®-2, Pore size: 90 Å mean 20 - 60 mesh ion-exchange resin, Supelco®
10357.
:
Merck
:
Pk:
1
Seq:
3645
Pk Info:
1× 500 g
:
:
9003-70-7