Amberlite® XAD4, Pore size: 100 Å 20 - 60 mesh ion-exchange resin, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:9003-69-4
:37380-42-0
MDL Number:MFCD00132704
:M-Clarity™ quality level = MQ100
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
VWR®, Tubing, Silicone
Supplier: VWR Collection
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals