Order Entry
Czech Republic
ContactUsLinkComponent
Oversleeves, embossed, CPE
Oversleeves, embossed, CPE
Catalog # HOIT115802
undefined
Oversleeves, embossed, CPE
Catalog # HOIT115802

Specifications

  • Colour:
    Blue
  • Length:
    440×220×0.03 m
  • Pk:
    1.000

Specifications

About this item

Weight: 7 g/m²

Packaging: 10 pcs/roll;10 rolls/polybag;10 bag/ carton(48×28×28cm)=1000 pcs