Order Entry
Czech Republic
ContactUsLinkComponent
Avantor® Hichrom, Connectors, Stainless Steel
Avantor® Hichrom, Connectors, Stainless Steel
Catalog # HICHHI-752
Supplier:  Avantor
undefined
Avantor® Hichrom, Connectors, Stainless Steel
Catalog # HICHHI-752
Supplier:  Avantor

Specifications

  • Description:
    ZDV male-male union for 1/16" tubing, 0,010" thru-hole
  • Material:
    Stainless steel
  • Seq:
    02
  • Pk:
    1

Specifications

About this item

The female-female connector consists of stainless steel tubing (40 mm×1,588×0,254 mm (L×O.Ø×I.Ø)) with appropriate 1/16" nuts and ferrules and can be used to connect female endfitting guard and analytical columns and make similar connections.

  • ZDV unions are rated to 20000 psi

The ZDV male-male connectors (unions) have internal threads and provide zero dead volume connections.