Order Entry
Czech Republic
ContactUsLinkComponent
Peqlab peqGOLD, DNase I Digest Kit
Peqlab peqGOLD, DNase I Digest Kit
Katalogové číslo 13-1091-00
Supplier:  VWR Chemicals
CAS Number:  
undefined
Peqlab peqGOLD, DNase I Digest Kit
Katalogové číslo 13-1091-00
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 Tests
  • Method / Format:
    Column (silica-based)
    Enzymatic clean-up
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Description:
    DNase I digest kit
  • Details:
    DNase I 10 μl, DNase digestion buffer 1 ml

Specifications

About this item

The peqGOLD DNase I Digest Kit is optimised for use with peqGOLD RNA extraction protocols. Normally DNase I digestion is not required for RNA purified with peqGOLD RNA Mini Columns as our silica-based spin column technology efficiently removes the majority of DNA without enzymatic digestion. However, certain sensitive RNA applications may require further DNA removal.

  • Efficient removal of contaminating DNA from RNA preparations
  • Free from RNases and inhibitors

Reaction time: 15 to 20 minutes on column at 20 to 30 °C
Activity: 10000 Kunitz units/mg
One Kunitz unit is defined as the amount of DNase I that causes an increase in A260 of 0,001 per minute per ml at 25 °C, pH 5,0, with highly polymerized DNA as the substrate.

Delivery: Kit contains RNase-free DNase I and DNase I digestion buffer.
Caution: Storage/stability: Store at −20 °C. The peqGOLD DNase I Digest Kit is stable for at least 12 months from the date of purchase when both DNase I and DNase Digestion Buffer are stored at −20 °C.