

2-Propanol, Semiconductor grade (Honeywell 17829)
Catalog # HONC40219H-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Boiling range:81 - 83 °C
- Density (D 20/4):0.785 - 0.787
- Refractive index (n 20/D):1.3770 - 1.3780
- Non-volatile matter:Max. 5 ppm
- Water (Karl Fischer):Max. 500 ppm
- Free acid (as C₂H₅COOH):Max. 10 ppm
- Silver (Ag):Max. 0.02 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.01 ppm
- Gold (Au):Max. 0.02 ppm
- Boron (B):Max. 0.01 ppm
- Barium (Ba):Max. 0.1 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.02 ppm
- Calcium (Ca):Max. 0.2 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Iron (Fe):Max. 0.1 ppm
- Gallium (Ga):Max. 0.02 ppm
- Indium (In):Max. 0.02 ppm
- Potassium (K):Max. 0.1 ppm
- Lithium (Li):Max. 0.02 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 0.2 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.02 ppm
- Platinum (Pt):Max. 0.02 ppm
- Antimony (Sb):Max. 0.01 ppm
- Tin (Sn):Max. 0.02 ppm
- Strontium (Sr):Max. 0.02 ppm
- Titanium (Ti):Max. 0.01 ppm
- Thallium (Tl):Max. 0.02 ppm
- Vanadium (V):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.01 ppm
- Chloride (Cl):Max. 1 ppm
- Sulfate (SO₄):Max. 1 ppm
- Acetone (GC):Max. 100 ppm
Specification Test Results
About this item
:
:60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674