Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
n-Pentane ≥95.0% (by GC), Laboratory Reagent
 
undefined
 
 
 
n-Pentane ≥95.0% (by GC), Laboratory Reagent

 

  • Assay (GC):
    Min. 95.0 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Non-volatile matter:
    Max. 0.002 %
  • Autoignition Temperature:
    500 °F
  • Refractive Index:
    n20/D 1.358 (lit.)
  • Vapor Density:
    2.48 (vs air)

 

 

Frequently Bought Together