Order Entry
Czech Republic
ContactUsLinkComponent
Toluene ≥99.5% (by GC), AR®, Macron Fine Chemicals™
Toluene ≥99.5% (by GC), AR®, Macron Fine Chemicals™
Certificates
undefined
Toluene ≥99.5% (by GC), AR®, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Assay (GC):
    ≥99.5%
  • Water (KF:
    ≤0.03 %
  • Titrable Acid (meq/g):
    ≤0.0005
  • Residue after Evaporation:
    ≤0.002 %
  • Sulfur Compounds:
    ≤0.0003 %

Specification Test Results

Related Information

Frequently Bought Together