



n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Catalog # 6752-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:86.18 g/mol
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520