0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Avantor® Hichrom, Unions
Avantor® Hichrom, Unions
Avantor® Hichrom, Unions
Catalog # HICHHI-089
Supplier:
Avantor
Expand image
Avantor® Hichrom, Unions
Catalog # HICHHI-089
Supplier:
Avantor
Specifications
Thread:
6-32
Description:
True ZDV union
Material:
PEEK
Seq:
01
Pk:
1
Specifications
About this item
Product Details
These unions are made of PEEK.
Product Details