Order Entry
Czech Republic
ContactUsLinkComponent
L-Glutamic acid, certified reference material, TraceCERT®, Supelco®
L-Glutamic acid, certified reference material, TraceCERT®, Supelco®
Catalog # 95436-100MG
Supplier:  Merck
CAS Number:  
undefined
L-Glutamic acid, certified reference material, TraceCERT®, Supelco®
Catalog # 95436-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    100 mg
  • Standards:
    Certified reference material
  • CAS No.:
    56-86-0
  • Description:
    L-Glutamic acid
  • Description:
    L-Glutamic acid
  • Pk Info:
    Bottomless glass bottle

Specifications

About this item

L-Glutamic acid has been used as a reference material in the development of quantitative nuclear magnetic resonance technique to evaluate the purity of organic molecules.

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.