ContactUsLinkComponent
LabSafe™ Nucleic acid stain (10,000X), G-Biosciences
  786-409
 :  
LabSafe™ Nucleic acid stain (10,000X), G-Biosciences
  786-409
 :  
  • Pk:
    1 mL
  • Description:
    LabSafe™ Nucleic Acid Stain (10,000X)
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels.
  • Non-carcinogenic by the Ames-test
  • Used commonly in gel electrophoresis for visualization of nucleic acids
  • Replace toxic Ethidium Bromide (a potent mutagen)
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels. The results are negative in both the mouse marrow chromophilous erythrocyte micronucleus and mouse primary spermatocyte chromosomal aberration tests.