Avantor® Alltima HP Cyano, HPLC Columns
Supplier: Avantor
Certificates
About this item
Avantor® Alltima HP Cyano HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated high purity silica phases
- Multiple selectivity options
- pH stability from 1 to 10
- Extremely stable, long life and reproducible
These columns are ideal for basic drug analysis. The low column bleed of these columns makes them as favourites for microbore applications.
Specifications
- Catalog No:
- HICH87769
- HICH87780
- 87784/N
- HICH87781
- HICH87760
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L10
- L10
- L10
- L10
- L10
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)