ContactUsLinkComponent
Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Catalog # P2548-100G
Supplier:  Merck
CAS Number:  
Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Catalog # P2548-100G
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 g
  • Description:
    Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Specifications
About this item
Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of heroin.
Caution: Storage temperature: −20 °C