Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    75-89-8
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226 - H301 + H331 - H318 - H360F - H373
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004672
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Trifluoroethyl alcohol
  • Melting point:
    –44 °C (lit.)
  • Formula:
    CF₃CH₂OH
  • Molecular weight:
    100,04
  • Density:
    1,373 g/ml at 25 °C (lit.)
  • Flash point:
    30 °C - Closed cup
  • Boiling point:
    77 - 80 °C (lit.)

Specifications

Related Information

Frequently Bought Together