Supelco® VETRANAL® Organic Reference Standard, Medroxyprogesterone 17-acetate
Catalog # 46412-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:VETRANAL®, analytical standard
- CAS No.:71-58-9
- Description:Medroxyprogesterone 17-acetate
- Storage conditions:2...30 °C
- Hazard class description:H351 - H413
- Shipping conditions:STD
- MDL No.:MFCD00010483
- M-Clarity quality level:MQ100
- Synonym(s):17α-Acetoxy-6α-methylprogesterone; 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate; 6α-Methyl-17α-acetoxyprogesterone; 6α-Methyl-17α-hydroxyprogesterone acetate
- Melting point:206 - 207 °C (lit.)
- Molecular weight:386,52
Specifications
About this item
- GHS signal: GHS08
Caution: P201 - P202 - P273 - P280 - P308 + P313 - P405