ContactUsLinkComponent
CytoRAL fixative for cytological smears, RAL
CytoRAL fixative for cytological smears, RAL
Catalog # 720-1153
Supplier:  RAL
CAS Number:  
CytoRAL fixative for cytological smears, RAL
Catalog # 720-1153
Supplier:  RAL
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Fixing spray 150 ml
Specifications
About this item
CytoRAL is used for cervical smears, urine, sputa and broncho-alveolar fluids.
It lays a protective film on the smears, keeping a good hydration before staining and without any alteration of the original cells aspect. With a large dispersion ratio the spray covers the entire slide area.