Order Entry
Czech Republic
ContactUsLinkComponent
Silver standard for ICP, TraceCERT®, Supelco®
Silver standard for ICP, TraceCERT®, Supelco®
Catalog # 12818-100ML
Supplier:  Merck
CAS Number:  
Silver standard for ICP, TraceCERT®, Supelco®
Catalog # 12818-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034