Order Entry
Czech Republic
ContactUsLinkComponent
Hanus solution (Iodine bromine) 0.1 mol/l (0.2 N), AVS TITRINORM for determination of iodine value
Hanus solution (Iodine bromine) 0.1 mol/l (0.2 N), AVS TITRINORM for determination of iodine value
Catalog # 86182.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hanus solution (Iodine bromine) 0.1 mol/l (0.2 N), AVS TITRINORM for determination of iodine value
Catalog # 86182.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Storage Temperature:Ambient
UN:2789
ADR:8,II
 :GHS02
 :GHS05
 :7789-33-5
Formula:IBr
MDL Number:MFCD00011353