ContactUsLinkComponent
HEMATOGNOST Fe® stain for histology, Sigma-Aldrich®
  1.12084.0001
 :  Merck
 :  
undefined
HEMATOGNOST Fe® stain for histology, Sigma-Aldrich®
  1.12084.0001
 :  Merck
 :  
  • Pack type:
    Fibre case
  • Pk:
    4
  • Seq:
    2596
  • Pk Info:
    4 x 250 ml
  • Potassium hexacyanoferrate II (Reagent 1):
    4.5 - 5.5 %
  • Hydrochloric acid (Reagent 2):
    4.5 - 5.5 %
  • Density (Reagent 3 d 20 °C/4 °C):
    1.040 - 1.060
  • Suitability for microscopy:
    passes test
 :
 :Ambient
 :3316
 :9,
 :GHS05
 :1