Order Entry
Czech Republic
ContactUsLinkComponent
RuBP-4S fluorescent dye, for histology
RuBP-4S fluorescent dye, for histology
Catalog # 80025.
Supplier:  Biotium
CAS Number:  
undefined
RuBP-4S fluorescent dye, for histology
Catalog # 80025.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1460.46 g/mol
Storage Temperature:Refrigerator
Formula:C₇₂H₄₄N₆Na₂O₁₂RuS₄