Order Entry
Czech Republic
ContactUsLinkComponent
Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt fluorescent dye, for histology
Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt fluorescent dye, for histology
Catalog # 80021.
Supplier:  Biotium
CAS Number:  
undefined
Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt fluorescent dye, for histology
Catalog # 80021.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1664.55 g/mol
Storage Temperature:Refrigerator
CAS Number:301206-84-8
Formula:C₇₂H₄₂N₆Na₄O₁₈RuS₆
MDL Number:MFCD04040034