Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Catalog # 32758-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Catalog # 32758-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    911392-43-3
  • Description:
    Zearalenone-13C18 solution
  • Formula:
    ₁₃C₁₈H₂₂O₅
  • Storage temperature:
    −20 °C
  • Description:
    Zearalenone-13C18 solution

Specifications

About this item

Zearalenone-¹³C₁₈ may be used as an analytical standard for the determination of zearalenone in animal feed by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

Zearalenone-¹³C₁₈ is the ¹³C isotope-labeled analog of the mycotoxin, zearalenone.