Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, T-2-Triol solution, analytical standard, Supelco®
Organic reference standard, T-2-Triol solution, analytical standard, Supelco®
Catalog # 32913-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, T-2-Triol solution, analytical standard, Supelco®
Catalog # 32913-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    34114-98-2
  • Description:
    T-2-Triol solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₀H₃₀O₇
  • Storage temperature:
    −20 °C
  • Description:
    T-2-Triol solution

Specifications

About this item

T-2-Triol is classified under the trichothecene family of mycotoxins that can adversely affect animal and human health. It is most commonly found in agricultural products.

T-2-Triol may be used as an analytical reference standard for the determination of the analyte in milk samples and layer feed samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS).