Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, Sterigmatocystin solution, analytical standard, Supelco®
Catalog # 32986-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sterigmatocystin solution, analytical standard, Supelco®
Catalog # 32986-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    10048-13-2
  • Description:
    Sterigmatocystin solution
  • Formula:
    C₁₈H₁₂O₆
  • Storage temperature:
    −20 °C
  • Description:
    Sterigmatocystin solution

Specifications

About this item

Sterigmatocystin is classified under the mycotoxin family of compounds. It is usually produced by the fungi of several species of Aspergillus.

Sterigmatocystin may be used as an analytical reference standard for the determination of the analyte in, Grain samples by high-performance liquid chromatography-electrospray positive ionization-tandem mass spectrometry (HPLC-ESI+ MS/MS). Beer samples by HPLC in conjunction with UV detection