Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Methyl 3-hydroxyhexanoate, analytical standard, Supelco®
Organic reference standard, Methyl 3-hydroxyhexanoate, analytical standard, Supelco®
Catalog # 77938-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Methyl 3-hydroxyhexanoate, analytical standard, Supelco®
Catalog # 77938-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    21188-58-9
  • Description:
    Methyl 3-hydroxyhexanoate, ≥98,0% (GC)
  • Formula:
    C₇H₁₄O₃
  • Description:
    Methyl 3-hydroxyhexanoate
  • Flash point:
    85 °C
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.