Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Menthone, analytical standard, Supelco®
Organic reference standard, Menthone, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Menthone, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    10458-14-7
  • Formula:
    C₁₀H₁₈O
  • Description:
    Menthone
  • Boiling point:
    85...88  °C/12 mm Hg (lit.)

Specifications

Frequently Bought Together