Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, ʟ-Proline hydrochloride solution
Catalog # 94321-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, ʟ-Proline hydrochloride solution
Catalog # 94321-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 mM amino acid in 0,1 M HCl
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    7776-34-3
  • Description:
    ʟ-Proline hydrochloride solution
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03791121
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    151,59

Specifications

About this item