Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Geranyl Pyrophosphate Lithium Salt
  19533-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Geranyl Pyrophosphate Lithium Salt
  19533-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    21141-43-5
  • Description:
    Geranyl pyrophosphate lithium salt
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (E)-3,7-Dimethyl-2,6-octadien-1-ol trihydrogen pyrophosphate lithium salt; (E)-3,7-Dimethyl-2,6-octadien-1-yl pyrophosphate lithium salt; GPP-Li
  • Formula:
    C₁₀H₂₀O₇P₂ ·xLi+
  • Molecular weight:
    314,21 (free acid basis)
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.