Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, D-ribonic acid lithium salt, ≥95,0% (TLC), Supelco®
 :  Merck
 
undefined
Organic reference standard, D-ribonic acid lithium salt, ≥95,0% (TLC), Supelco®
 :  Merck

 

  • CAS No.:
    487-06-9
  • Melting point:
    146...149 °C (lit.)
  • Formula:
    C₁₁H₁₀O₄
  • Description:
    D-Ribonic acid lithium salt

 

Frequently Bought Together