Organic reference standard, Diallyl disulfide, analytical standard, Supelco®
30648-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:2179-57-9
- Description:Diallyl disulfide, ≥96,0% (GC)
- Formula:C₆H₁₀S₂
- Storage temperature:2...8 °C
- Description:Diallyl disulfide