Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, D-2-Phosphoglyceric Acid Lithium Salt
Supelco® Organic Reference Standard, D-2-Phosphoglyceric Acid Lithium Salt
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, D-2-Phosphoglyceric Acid Lithium Salt
Supplier:  Merck

Specifications

  • CAS No.:
    3443-57-0
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2R)-3-Hydroxy-2-(phosphonooxy)propanoic acid lithium salt; Lithium ᴅ-2-phosphoglycerate
  • Formula:
    C₃H₇O₇P·xLi+
  • Molecular weight:
    186,06 (free acid basis)
  • Storage temperature:
    −20°C
  • Flash point:
    Not applicable

Specifications

Frequently Bought Together