Supelco® Organic Reference Standard, Cysteamine hydrochloride
49705-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:156-57-0
- Description:Cysteamine hydrochloride
- Storage conditions:2…8 °C
- Hazard class description:H302 - H317 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00012904
- M-Clarity quality level:MQ100
- Synonym(s):β-Mercaptoethylamine hydrochloride; 2-Aminoethanethiol hydrochloride; 2-Mercaptoethylamine hydrochloride; Decarboxycysteine hydrochloride; Thioethanolamine hydrochloride
- Melting point:67 - 71 °C
- Formula:HSCH₂CH₂NH₂ · HCl
- Molecular weight:113,61
- GHS signal: GHS07
: P261 - P264 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338