Phytochemical standard, cyanidin 3-sambubioside chloride, analytical standard, Supelco®
43046-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:33012-73-6
- Description:Cyanidin 3-sambubioside chloride, ≥95,0% (HPLC)
- Formula:C₂₆H₂₉ClO₁₅
- Storage temperature:–20 °C
- Description:Cyanidin 3-sambubioside chloride