Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Citrinin solution, analytical standard, Supelco®
Organic reference standard, Citrinin solution, analytical standard, Supelco®
Catalog # 35410-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Citrinin solution, analytical standard, Supelco®
Catalog # 35410-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    518-75-2
  • Description:
    Citrinin solution
  • Formula:
    C₁₃H₁₄O₅
  • Storage temperature:
    −20 °C
  • Description:
    Citrinin solution

Specifications

About this item

Citrinin is a hepato-nephrotoxic mycotoxin. It can be produced by several filamentous fungi genera of Monauscus, Aspergillus, and Penicillium, which is commonly found as a contaminant in stored grains, foods, feedstuffs and also in biological fluids.