Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, Aflatoxin G1-13C17 solution, analytical standard, Supelco®
Catalog # 32772-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aflatoxin G1-13C17 solution, analytical standard, Supelco®
Catalog # 32772-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.5 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1217444-07-9
  • Description:
    Aflatoxin G1-13C17 solution
  • Formula:
    13C₁₇H₁₂O₇
  • Storage temperature:
    –20 °C
  • Description:
    Aflatoxin G1-13C17 solution

Specifications

About this item

Aflatoxin G1-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.

It may be used as an analytical standard for the determination of aflatoxin G1 in food grade gums as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).