Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Aflatoxin B1 solution, analytical standard, Supelco®
Organic reference standard, Aflatoxin B1 solution, analytical standard, Supelco®
Catalog # 34029-2ML-R
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aflatoxin B1 solution, analytical standard, Supelco®
Catalog # 34029-2ML-R
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1162-65-8
  • Description:
    Aflatoxin B₁ solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₇H₁₂O₆
  • Storage temperature:
    −20 °C
  • Description:
    Aflatoxin B₁ solution
  • Flash point:
    2 °C

Specifications

About this item

Aflatoxin B1 is a mycotoxin found to be hepatocarcinogenic in nature. It has a strong affinity for the deoxyribonucleic acid (DNA) and binds to both native and denatured deoxyribonucleic acid (DNA) as indicated by spectroscopy and equilibrium dialysis results.