Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Acid Red 73, analytical standard, Supelco®
Organic reference standard, Acid Red 73, analytical standard, Supelco®
Katalogové číslo 49823-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Acid Red 73, analytical standard, Supelco®
Katalogové číslo 49823-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5413-75-2
  • Description:
    Acid Red 73, ≥97,0% (HPLC)
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₂H₁₄N₄Na₂O₇S₂
  • Description:
    Acid Red 73
  • Form:
    Neat

Specifications

About this item

Acid Red 73 is a synthetically prepared sulfonated azo dye.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.