Supelco® Organic Reference Standard, Acetyleugenol
Catalog # 04733-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:93-28-7
- Description:Acetyleugenol
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H302 - H317
- Shipping conditions:STD
- MDL No.:MFCD00026191
- M-Clarity quality level:MQ100
- Synonym(s):Eugenyl acetate; 4-Allyl-2-methoxyphenyl acetate; O-Acetyleugenol; Eugenol acetate
- Formula:C₁₂H₁₄O₃
- Molecular weight:206,24
- Density:1,079 g/ml at 25 °C (lit.)
- Flash point:142 °C - closed cup
- Boiling point:281...286 °C (lit.)
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P270 - P280 - P301 + P312 - P302 + P352