Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, 14-Deoxy-11,12-didehydroandrographolide
Supelco® Organic Reference Standard, 14-Deoxy-11,12-didehydroandrographolide
Catalog # 55549-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 14-Deoxy-11,12-didehydroandrographolide
Catalog # 55549-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    42895-58-9
  • Description:
    14-Deoxy-11,12-didehydroandrographolide
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07778081
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-[(1E)-2-[(1R,4aS,5R,6R,8aR)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethenyl]- 2(5H)-furanone; ent-(3β,11E)-3,19-Dihydroxy-8(17),11,13-labdatrien-16,15-olide
  • Molecular weight:
    332,43

Specifications

About this item