Supelco® Organic Reference Standard, 11-Keto-β-boswellic acid
78535-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:17019-92-0
- Description:11-Keto-β-boswellic acid
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD06656312
- M-Clarity quality level:MQ100
- Synonym(s):11-Oxo-β-boswellic acid; 3α-Hydroxy-11-oxours-12-en-24-oic acid; KBA
- Molecular weight:470,68