Phytochemical standard, 1,5-Dicaffeoylquinic acid, analytical standard, Supelco®
16917-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:19870-46-3
- Description:1,5-Dicaffeoylquinic acid, ≥98,0% (HPLC)
- Formula:C₂₅H₂₄O₁₂
- Storage temperature:2…8 °C
- Description:1,5-Dicaffeoylquinic acid
- Form:Neat
Dicaffeoylquinic acid is naturally occurring polyphenolic compound found in plants like fennel and coffee. It is characterized by two caffeic acid molecules linked via ester bonds to one quinic acid molecule. 1, 5-Dicaffeoylquinic acid (1,5-DCQA) is considered as a potent and nontoxic inhibitor of HIV-1 replication.
Biochemical and physiol actions are phenolic compound found in artichoke. Antioxidant and choleretic properties. Potential immunosuppressive agent.
: Bottomless glass bottle. Contents are inside inserted fused cone.