Organic reference standard, (+/-)-alpha-Terpinyl acetate, predominantly alpha-isomer, analytical standard, Supelco®
Katalogové číslo 09206-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:80-26-2
- Description:(+/-)-alpha-Terpinyl acetate, predominantly alpha-isomer, ≤15% (γ-isomer, GC), ≥85% (α-isomer, GC), ≥97,0% (sum of isomers, GC)
- Formula:C₁₂H₂₀O₂
- Description:(+/-)-alpha-Terpinyl acetate
- Boiling point:220 °C (lit.)