Order Entry
Czech Republic
ContactUsLinkComponent
Lucifer yellow CH dilithium salt fluorescent dye, for histology
Lucifer yellow CH dilithium salt fluorescent dye, for histology
Catalog # 80015.
Supplier:  Biotium
CAS Number:  
undefined
Lucifer yellow CH dilithium salt fluorescent dye, for histology
Catalog # 80015.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :457.25 g/mol
Storage Temperature:Refrigerator
EINECS:267-047-9
CAS Number:67769-47-5
Formula:C₁₃H₉N₅O₉S₂Li₂
MDL Number:MFCD00006922